Product Details

SNP ID
rs202044839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176152852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAAGGCAGGTCATCGTCCTCATC[C/T]TCCTCCTCATCTTGCTCCTCCTCAG
Phenotype
MIM: 142980 MIM: 142981 MIM: 611576
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXD3 PubMed Links

Gene Details

Gene
HOXD3
Gene Name
homeobox D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006898.4 1821 Intron NP_008829.3
XM_005246509.3 1821 Intron XP_005246566.1
XM_005246510.4 1821 Intron XP_005246567.1
XM_005246511.3 1821 Intron XP_005246568.1
XM_005246513.4 1821 Intron XP_005246570.1
XM_006712477.2 1821 Intron XP_006712540.1
XM_011511065.2 1821 Intron XP_011509367.1
XM_011511066.2 1821 Intron XP_011509368.1
Gene
HOXD4
Gene Name
homeobox D4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014621.2 1821 Silent Mutation TCC,TCT S226S NP_055436.2
XM_005246514.4 1821 UTR 3 XP_005246571.1
Gene
MIR10B
Gene Name
microRNA 10b
There are no transcripts associated with this gene.

View Full Product Details