Product Details

SNP ID
rs1131553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184389064 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCCTCTTCCGCCAGTGTGGCCG[T/C]CCCTTTCAAGAAAGTAATTAAGATA
Phenotype
MIM: 147576
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
IRF2 PubMed Links

Gene Details

Gene
IRF2
Gene Name
interferon regulatory factor 2
There are no transcripts associated with this gene.

Gene
LOC102723766
Gene Name
uncharacterized LOC102723766
There are no transcripts associated with this gene.

View Full Product Details