Product Details

SNP ID
rs200052188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9934465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCCCATGGCCCCCGAAGGGCCT[A/G]GTCCCAGCTATGCTCTGGGGCCTCA
Phenotype
MIM: 607170 MIM: 610925
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRELD1 PubMed Links

Gene Details

Gene
CRELD1
Gene Name
cysteine rich with EGF like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031717.3 126 Silent Mutation CTA,CTG L9L NP_001026887.1
NM_001077415.2 126 Silent Mutation CTA,CTG L9L NP_001070883.1
NM_015513.4 126 Silent Mutation CTA,CTG L9L NP_056328.2
XM_011534108.1 126 Silent Mutation CTA,CTG L9L XP_011532410.1
XM_017007175.1 126 Silent Mutation CTA,CTG L9L XP_016862664.1
Gene
IL17RC
Gene Name
interleukin 17 receptor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203263.1 126 Intron NP_001190192.1
NM_001203264.1 126 Intron NP_001190193.1
NM_001203265.1 126 Intron NP_001190194.1
NM_032732.5 126 Intron NP_116121.2
NM_153460.3 126 Intron NP_703190.1
NM_153461.3 126 Intron NP_703191.1
XM_017007326.1 126 Intron XP_016862815.1
XM_017007327.1 126 Intron XP_016862816.1
XM_017007328.1 126 Intron XP_016862817.1
XM_017007329.1 126 Intron XP_016862818.1
XM_017007330.1 126 Intron XP_016862819.1
XM_017007331.1 126 Intron XP_016862820.1
XM_017007332.1 126 Intron XP_016862821.1
XM_017007333.1 126 Intron XP_016862822.1
XM_017007334.1 126 Intron XP_016862823.1
XM_017007335.1 126 Intron XP_016862824.1
XM_017007336.1 126 Intron XP_016862825.1
XM_017007337.1 126 Intron XP_016862826.1
XM_017007338.1 126 Intron XP_016862827.1
XM_017007339.1 126 Intron XP_016862828.1
XM_017007340.1 126 Intron XP_016862829.1
XM_017007341.1 126 Intron XP_016862830.1
XM_017007342.1 126 Intron XP_016862831.1
XM_017007343.1 126 Intron XP_016862832.1
XM_017007344.1 126 Intron XP_016862833.1
XM_017007345.1 126 Intron XP_016862834.1
XM_017007346.1 126 Intron XP_016862835.1
XM_017007347.1 126 Intron XP_016862836.1
XM_017007348.1 126 Intron XP_016862837.1
XM_017007349.1 126 Intron XP_016862838.1

View Full Product Details