Product Details

SNP ID
rs201668251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73854037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTTCTCTTTTCCCTGCCAGAG[C/T]CACGCTGAAGAATGGGAGGAAAATT
Phenotype
MIM: 173461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PF4V1 PubMed Links

Gene Details

Gene
PF4V1
Gene Name
platelet factor 4 variant 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002620.3 297 Missense Mutation GCC,GTC A77V NP_002611.1

View Full Product Details