Product Details

SNP ID
rs200449617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140632619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGTTATCTTTAGGTCCTCGAGC[A/G]TCAGTTCCTTGAGGCGGGAGTACGC
Phenotype
MIM: 158120
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD14 PubMed Links

Gene Details

Gene
CD14
Gene Name
CD14 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000591.3 757 Missense Mutation ACG,ATG T122M NP_000582.1
NM_001040021.2 757 Missense Mutation ACG,ATG T122M NP_001035110.1
NM_001174104.1 757 Missense Mutation ACG,ATG T122M NP_001167575.1
NM_001174105.1 757 Missense Mutation ACG,ATG T122M NP_001167576.1
Gene
TMCO6
Gene Name
transmembrane and coiled-coil domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300980.1 757 Intron NP_001287909.1
NM_001300982.1 757 Intron NP_001287911.1
NM_018502.4 757 Intron NP_060972.3
XM_005268477.1 757 Intron XP_005268534.1
XM_011537663.2 757 Intron XP_011535965.1
XM_011537665.2 757 Intron XP_011535967.1
XM_011537668.2 757 Intron XP_011535970.1
XM_017009617.1 757 Intron XP_016865106.1
XM_017009618.1 757 Intron XP_016865107.1
XM_017009619.1 757 Intron XP_016865108.1
XM_017009620.1 757 Intron XP_016865109.1

View Full Product Details