Product Details

SNP ID
rs201094763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:105112271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACTTTTATCCAGCAGCACAGAA[C/T]CTTCCAGCCAATACTCAGCAGGCAA
Phenotype
MIM: 608444 MIM: 602980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KMT2E PubMed Links

Gene Details

Gene
KMT2E
Gene Name
lysine methyltransferase 2E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018682.3 4968 Silent Mutation AAC,AAT N1505N NP_061152.3
NM_182931.2 4968 Silent Mutation AAC,AAT N1505N NP_891847.1
XM_005250493.1 4968 Silent Mutation AAC,AAT N1505N XP_005250550.1
XM_011516400.1 4968 Silent Mutation AAC,AAT N1505N XP_011514702.1
XM_017012435.1 4968 Silent Mutation AAC,AAT N1359N XP_016867924.1
Gene
SRPK2
Gene Name
SRSF protein kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278273.1 4968 Intron NP_001265202.1
NM_182691.2 4968 Intron NP_872633.1
NM_182692.2 4968 Intron NP_872634.1
XM_005250549.3 4968 Intron XP_005250606.2
XM_005250550.3 4968 Intron XP_005250607.1
XM_005250551.4 4968 Intron XP_005250608.1
XM_006716098.2 4968 Intron XP_006716161.1
XM_011516536.2 4968 Intron XP_011514838.1
XM_011516538.2 4968 Intron XP_011514840.1
XM_017012560.1 4968 Intron XP_016868049.1
XM_017012561.1 4968 Intron XP_016868050.1
XM_017012562.1 4968 Intron XP_016868051.1
XM_017012563.1 4968 Intron XP_016868052.1
XM_017012564.1 4968 Intron XP_016868053.1
XM_017012565.1 4968 Intron XP_016868054.1
XM_017012566.1 4968 Intron XP_016868055.1

View Full Product Details