Product Details

SNP ID
rs201839000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:90625717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGAAAATGTTAGGGTCCTCTTG[A/T]TGTAGAATGAAGAGCCACTGGTATT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM64 PubMed Links

Gene Details

Gene
TMEM64
Gene Name
transmembrane protein 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008495.3 1097 Missense Mutation AAC,ATC N366I NP_001008495.2
NM_001146273.1 1097 Missense Mutation AAC,ATC N314I NP_001139745.1

View Full Product Details