Product Details

SNP ID
rs200627906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:76392476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCCTATGCTCACCACCATCTTA[C/T]GGACATCTCCACTTCCAACACTGGC
Phenotype
MIM: 613010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RFK PubMed Links

Gene Details

Gene
RFK
Gene Name
riboflavin kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018339.5 533 Missense Mutation CAT,CGT H59R NP_060809.3
Gene
RPSAP9
Gene Name
ribosomal protein SA pseudogene 9
There are no transcripts associated with this gene.

View Full Product Details