Product Details
- SNP ID
-
rs200773334
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:89325964 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGACAGTGAAAGGGCTGATGGAA[C/T]TATATCATCTGAGATAAAATCAGCT
- Phenotype
-
MIM: 116901
MIM: 607693
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CKS2
PubMed Links
Gene Details
- Gene
- CKS2
- Gene Name
- CDC28 protein kinase regulatory subunit 2
There are no transcripts associated with this gene.
- Gene
- SECISBP2
- Gene Name
- SECIS binding protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282688.1 |
658 |
Missense Mutation |
ACT,ATT |
T166I |
NP_001269617.1 |
NM_001282689.1 |
658 |
Missense Mutation |
ACT,ATT |
T94I |
NP_001269618.1 |
NM_001282690.1 |
658 |
Missense Mutation |
ACT,ATT |
T99I |
NP_001269619.1 |
NM_024077.4 |
658 |
Missense Mutation |
ACT,ATT |
T167I |
NP_076982.3 |
XM_005252196.2 |
658 |
Missense Mutation |
ACT,ATT |
T167I |
XP_005252253.1 |
XM_005252202.3 |
658 |
Intron |
|
|
XP_005252259.1 |
XM_006717282.2 |
658 |
Missense Mutation |
ACT,ATT |
T94I |
XP_006717345.1 |
XM_011519000.2 |
658 |
Missense Mutation |
ACT,ATT |
T99I |
XP_011517302.1 |
XM_011519001.1 |
658 |
Missense Mutation |
ACT,ATT |
T99I |
XP_011517303.1 |
XM_011519002.1 |
658 |
Missense Mutation |
ACT,ATT |
T99I |
XP_011517304.1 |
XM_011519003.1 |
658 |
Missense Mutation |
ACT,ATT |
T99I |
XP_011517305.1 |
XM_017015122.1 |
658 |
Missense Mutation |
ACT,ATT |
T167I |
XP_016870611.1 |
XM_017015123.1 |
658 |
Missense Mutation |
ACT,ATT |
T167I |
XP_016870612.1 |
XM_017015124.1 |
658 |
Missense Mutation |
ACT,ATT |
T167I |
XP_016870613.1 |
XM_017015125.1 |
658 |
UTR 5 |
|
|
XP_016870614.1 |
View Full Product Details