Product Details

SNP ID
rs200322025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49191579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTAGCCGCCCTGAGGCCCGTAG[A/G]AATCCTGGGGCCCGTACCCGCCGGG
Phenotype
MIM: 300111 MIM: 313475
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRICKLE3 PubMed Links

Gene Details

Gene
PRICKLE3
Gene Name
prickle planar cell polarity protein 3
There are no transcripts associated with this gene.

Gene
SYP
Gene Name
synaptophysin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003179.2 816 Missense Mutation TCC,TTC S267F NP_003170.1
Gene
SYP-AS1
Gene Name
SYP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details