Product Details
- SNP ID
-
rs2242607
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:73242476 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTGACAACCAGGCCCAACCCAGCAG[C/T]TAAGAAAGCAGCCACCCCTGAAGAC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101928123
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116826539] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101928123
- Gene Name
- uncharacterized LOC101928123
There are no transcripts associated with this gene.
- Gene
- PAPLN
- Gene Name
- papilin, proteoglycan like sulfated glycoprotein
View Full Product Details