Product Details

SNP ID
rs12794028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3221458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACCACTGCGTCTGTGGGTGGTCT[A/G]CAGGGGCGGCCCCCACCCCTTCCTA
Phenotype
MIM: 607232 MIM: 607234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRGPRE PubMed Links

Gene Details

Gene
MRGPRE
Gene Name
MAS related GPR family member E
There are no transcripts associated with this gene.

Gene
MRGPRG
Gene Name
MAS related GPR family member G
There are no transcripts associated with this gene.

Gene
MRGPRG-AS1
Gene Name
MRGPRG antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details