Product Details

SNP ID
rs1128665
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:36339711 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATGTCTTTATACAAACTTTTTTG[G/T]GACTTTTTCCGTTTCTTTACAATAG
Phenotype
MIM: 609437
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
STK40 PubMed Links

Gene Details

Gene
STK40
Gene Name
serine/threonine kinase 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282546.1 3802 UTR 3 NP_001269475.1
NM_001282547.1 3802 UTR 3 NP_001269476.1
NM_032017.2 3802 UTR 3 NP_114406.1

View Full Product Details