Product Details
- SNP ID
-
rs1811257
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:4967386 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGCCTCGCTTCAACTTGATTACTT[A/G]TAAAGACCTGTTCTCCAAATGAGGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEC14L5
PubMed Links
Gene Details
- Gene
- SEC14L5
- Gene Name
- SEC14 like lipid binding 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014692.1 |
|
Intron |
|
|
NP_055507.1 |
View Full Product Details