Product Details

SNP ID
rs2645081
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:2397275 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCTGTCCACGTTACCTGGGCGTG[G/A]TGTTTGTGGGGATGTTGTTGATCCA
Phenotype
MIM: 602859
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MORN1 PubMed Links

Gene Details

Gene
MORN1
Gene Name
MORN repeat containing 1
There are no transcripts associated with this gene.

Gene
PEX10
Gene Name
peroxisomal biogenesis factor 10
There are no transcripts associated with this gene.

Gene
RER1
Gene Name
retention in endoplasmic reticulum sorting receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007033.4 Intron NP_008964.3
XM_005244713.4 Intron XP_005244770.2
XM_006710306.2 Intron XP_006710369.1
XM_011540542.1 Intron XP_011538844.1
XM_011540543.1 Intron XP_011538845.1
XM_017000131.1 Intron XP_016855620.1
XM_017000132.1 Intron XP_016855621.1
XM_017000133.1 Intron XP_016855622.1
XM_017000134.1 Intron XP_016855623.1
XM_017000135.1 Intron XP_016855624.1

View Full Product Details