Product Details

SNP ID
rs1976332
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:66800163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATTTTTAAGTTAGTTGAATTGA[A/G]TAATAAAGAAATATAGTGAAGTCAG
Phenotype
MIM: 606413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INSL5 PubMed Links

Gene Details

Gene
INSL5
Gene Name
insulin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005478.4 Intron NP_005469.2

View Full Product Details