Product Details

SNP ID
rs1816424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:13940879 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTGGTGACTGTTGTGCCTTTTAA[A/T]ACATAGAGGTGCTTTGCCTGATACC
Phenotype
MIM: 611167
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FGD5P1 PubMed Links

Gene Details

Gene
FGD5P1
Gene Name
FYVE, RhoGEF and PH domain containing 5 pseudogene 1
There are no transcripts associated with this gene.

Gene
TPRXL
Gene Name
tetrapeptide repeat homeobox like
There are no transcripts associated with this gene.

View Full Product Details