Product Details

SNP ID
rs11045522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:20812510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATTCCAGGAATGGAAGACATA[C/T]ACATGAAGGTAGAGAGCTTTTCTGT
Phenotype
MIM: 605495
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLCO1B3 PubMed Links

Gene Details

Gene
SLCO1B3
Gene Name
solute carrier organic anion transporter family member 1B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019844.3 Intron NP_062818.1

View Full Product Details