Product Details

SNP ID
rs2296323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:65004468 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTACCTGGATATGCTAAGACCCC[C/T]ACTCCTCCCTCTGGTAGAATACTGT
Phenotype
MIM: 134636 MIM: 154950
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHURC1-FNTB PubMed Links

Gene Details

Gene
CHURC1-FNTB
Gene Name
CHURC1-FNTB readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202558.1 Intron NP_001189487.1
NM_001202559.1 Intron NP_001189488.1
Gene
FNTB
Gene Name
farnesyltransferase, CAAX box, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002028.3 Intron NP_002019.1
Gene
MAX
Gene Name
MYC associated factor X
There are no transcripts associated with this gene.

View Full Product Details