Product Details
- SNP ID
-
rs2384384
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:26360765 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGTGAAGTAAAGGAGTGTAGGGC[C/T]CTCCTATCTTGTCATTGGCAGCCTG
- Phenotype
-
MIM: 607915
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPT1
PubMed Links
Gene Details
- Gene
- EPT1
- Gene Name
- ethanolaminephosphotransferase 1
There are no transcripts associated with this gene.
View Full Product Details