Product Details

SNP ID
rs2421456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34058513 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGTGTAGTTTTCATGTTTAATTA[C/T]GTAGAATCAAGTAAAGCCTTTTGCT
Phenotype
MIM: 601178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPRIN1 PubMed Links

Gene Details

Gene
CAPRIN1
Gene Name
cell cycle associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005898.4 Intron NP_005889.3
NM_203364.2 Intron NP_976240.1
XM_017017749.1 Intron XP_016873238.1

View Full Product Details