Product Details

SNP ID
rs2075966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111563565 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGAAAAGTATTAACAATTGCTCT[A/G]GAAGCCCAGAATGGGGGCCATCCAT
Phenotype
MIM: 607043
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAF3IP2 PubMed Links

Gene Details

Gene
TRAF3IP2
Gene Name
TRAF3 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164281.2 Intron NP_001157753.1
NM_001164283.2 Intron NP_001157755.1
NM_147200.2 Intron NP_671733.2
NM_147686.3 Intron NP_679211.2
XM_006715319.3 Intron XP_006715382.1
XM_011535386.1 Intron XP_011533688.1
Gene
TRAF3IP2-AS1
Gene Name
TRAF3IP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details