Product Details

SNP ID
rs2288946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5562073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTCTCTTTCTCAACTACTACA[A/G]TTACTTACCTGGAGTGAGATCTAGG
Phenotype
MIM: 615241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TINCR PubMed Links

Gene Details

Gene
TINCR
Gene Name
tissue differentiation-inducing non-protein coding RNA
There are no transcripts associated with this gene.

View Full Product Details