Product Details

SNP ID
rs2683003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:118356300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCACTGCATTTCAAAGAGTTATG[G/T]TCTTTGTTCCTAAGGGAAACCCCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WDR44 PubMed Links

Gene Details

Gene
WDR44
Gene Name
WD repeat domain 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184965.1 Intron NP_001171894.1
NM_001184966.1 Intron NP_001171895.1
NM_019045.4 Intron NP_061918.3
XM_011531353.2 Intron XP_011529655.1
XM_017029600.1 Intron XP_016885089.1

View Full Product Details