Product Details

SNP ID
rs2721121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18456310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGAGAACTGGAGGTTGGGGGAA[A/C]AGCAGGGTAAAGGGGAGAGAAAAGG
Phenotype
MIM: 150150
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LDHAL6A PubMed Links

Gene Details

Gene
LDHAL6A
Gene Name
lactate dehydrogenase A like 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144071.1 74 Intron NP_001137543.1
NM_144972.4 74 UTR 5 NP_659409.2
XM_005252805.4 74 Intron XP_005252862.2
XM_011519922.2 74 Intron XP_011518224.1
XM_011519923.2 74 Intron XP_011518225.1
XM_011519924.2 74 UTR 5 XP_011518226.1
XM_011519925.1 74 Intron XP_011518227.1
Gene
LDHC
Gene Name
lactate dehydrogenase C
There are no transcripts associated with this gene.

View Full Product Details