Product Details
- SNP ID
-
rs2961160
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:144129510 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCTGCCACCCCTTACGTTACAATA[G/T]CCTCATGAGCTGGAGAGTGTGCACT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2A14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2961161] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2A14
- Gene Name
- olfactory receptor family 2 subfamily A member 14
View Full Product Details