Product Details

SNP ID
rs2982499
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:40450197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGTGAGCGGGTGGCTGTCCCGG[A/G]CTTGGGGAGCTGGGCGAGCCGGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFP69B PubMed Links

Gene Details

Gene
ZFP69B
Gene Name
ZFP69 zinc finger protein B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023070.2 Intron NP_075558.2
XM_005271136.1 Intron XP_005271193.1
XM_005271137.2 Intron XP_005271194.1
XM_005271138.3 Intron XP_005271195.1
XM_005271139.3 Intron XP_005271196.1
XM_017002147.1 Intron XP_016857636.1

View Full Product Details