Product Details

SNP ID
rs2291877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140118549 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAACCAGAACTGTAGAAATCATTA[C/T]ACATGCCTTTGACAACAAAGGAAGT
Phenotype
MIM: 600473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGIP PubMed Links

Gene Details

Gene
IGIP
Gene Name
IgA inducing protein
There are no transcripts associated with this gene.

Gene
PURA
Gene Name
purine rich element binding protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005859.4 4427 UTR 3 NP_005850.1

View Full Product Details