Product Details

SNP ID
rs2304467
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:8894920 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCACCCCCAGGCCACGTCACGTGG[C/G]AGCCGCCAAAACCAACGCCTAACCC
Phenotype
MIM: 602519
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929989 PubMed Links

Gene Details

Gene
LOC101929989
Gene Name
lipopolysaccharide-induced tumor necrosis factor-alpha factor-like
There are no transcripts associated with this gene.

Gene
USP7
Gene Name
ubiquitin specific peptidase 7 (herpes virus-associated)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286457.1 Intron NP_001273386.1
NM_001286458.1 Intron NP_001273387.1
NM_001321858.1 Intron NP_001308787.1
NM_003470.2 Intron NP_003461.2
XM_017023652.1 Intron XP_016879141.1
XM_017023653.1 Intron XP_016879142.1

View Full Product Details