Product Details

SNP ID
rs2863437
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:16044169 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGACCCAGGAGGGTGGGAGCGGC[G/T]GCCACCGAAGGGGAAGGGGCTCTGA
Phenotype
MIM: 602023
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLCNKB PubMed Links

Gene Details

Gene
CLCNKB
Gene Name
chloride voltage-gated channel Kb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000085.4 Intron NP_000076.2
NM_001165945.2 Intron NP_001159417.2

View Full Product Details