Product Details

SNP ID
rs2107547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58355382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCAGTGGGACAGGACAGGAGTCA[C/T]GAGAGATGCATTAAGGAAGGAGAGA
Phenotype
MIM: 612482 MIM: 603555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF43 PubMed Links

Gene Details

Gene
RNF43
Gene Name
ring finger protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305544.1 Intron NP_001292473.1
NM_001305545.1 Intron NP_001292474.1
NM_017763.5 Intron NP_060233.3
XM_011524955.2 Intron XP_011523257.1
XM_011524956.2 Intron XP_011523258.1
XM_017024800.1 Intron XP_016880289.1
Gene
SUPT4H1
Gene Name
SPT4 homolog, DSIF elongation factor subunit
There are no transcripts associated with this gene.

Gene
TSPOAP1-AS1
Gene Name
TSPOAP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details