Product Details

SNP ID
rs2298214
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:999842 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
GTCCCTCCCGCCCCCCGGTCGCCCC[A/C]CTCACGCCCGGCCGGGACCCCACCT
Phenotype
MIM: 608060
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HES4 PubMed Links

Gene Details

Gene
HES4
Gene Name
hes family bHLH transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142467.1 331 Missense Mutation AGG,AGT R44S NP_001135939.1
NM_021170.3 331 Intron NP_066993.1
XM_005244771.4 331 Intron XP_005244828.1

View Full Product Details