Product Details
- SNP ID
-
rs2306586
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:36546846 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTTTGTTCCTCAATATTTTTTTT[G/T]GCAAATATCTACCGAAGTTTTTTCA
- Phenotype
-
MIM: 612275
MIM: 610275
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GGNBP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74479687] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GGNBP2
- Gene Name
- gametogenetin binding protein 2
- Gene
- PIGW
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class W
There are no transcripts associated with this gene.
View Full Product Details