Product Details

SNP ID
rs2576692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55289587 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCAAGATTTTCAAAATGATTTAA[C/T]GTTAACAGCATTTGGTGGTAAAACC
Phenotype
MIM: 609736
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC88A PubMed Links

Gene Details

Gene
CCDC88A
Gene Name
coiled-coil domain containing 88A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135597.1 7693 UTR 3 NP_001129069.1
NM_001254943.1 7693 UTR 3 NP_001241872.1
NM_018084.4 7693 UTR 3 NP_060554.3
XM_005264418.4 7693 Intron XP_005264475.1
XM_005264421.2 7693 UTR 3 XP_005264478.1
XM_005264426.2 7693 Intron XP_005264483.1
XM_011532965.2 7693 UTR 3 XP_011531267.1
XM_011532966.2 7693 Intron XP_011531268.1
XM_011532967.2 7693 Intron XP_011531269.1
XM_011532968.2 7693 UTR 3 XP_011531270.1
XM_017004476.1 7693 Intron XP_016859965.1
XM_017004477.1 7693 Intron XP_016859966.1
XM_017004478.1 7693 Intron XP_016859967.1
XM_017004479.1 7693 Intron XP_016859968.1
XM_017004480.1 7693 Intron XP_016859969.1
XM_017004481.1 7693 Intron XP_016859970.1
Gene
PRORSD1P
Gene Name
prolyl-tRNA synthetase associated domain containing 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details