Product Details

SNP ID
rs2735682
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:4736225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATCCATTCCCAATGTTTTCATTC[A/T]CGTGACAATTCTTCTTGGTCTCAGT
Phenotype
MIM: 604263
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
PRND PubMed Links
Additional Information
For this assay, SNP(s) [rs75611931] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PRND
Gene Name
prion protein 2 (dublet)
There are no transcripts associated with this gene.

Gene
PRNT
Gene Name
prion protein (testis specific)
There are no transcripts associated with this gene.

View Full Product Details