Product Details
- SNP ID
-
rs2757589
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:163422186 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTTACAATTAGCAAGAAAAGGAC[C/T]GCACTTAATGTTTTCGTCTTCAGTT
- Phenotype
-
MIM: 615930
MIM: 609590
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CAHM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150200739] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAHM
- Gene Name
- colon adenocarcinoma hypermethylated (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- QKI
- Gene Name
- QKI, KH domain containing RNA binding
View Full Product Details