Product Details
- SNP ID
-
rs61735970
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:100721973 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGTGCATTTCAGACGGGCTGTGC[C/T]GAACACTGCAGCTTGAATGAGAATA
- Phenotype
-
MIM: 133170
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPO
PubMed Links
Gene Details
- Gene
- EPO
- Gene Name
- erythropoietin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000799.2 |
352 |
Silent Mutation |
GCC,GCT |
A57A |
NP_000790.2 |
View Full Product Details