Product Details

SNP ID
rs61736596
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:68046758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACATTCACATCGGTCTTGGCTGA[A/G]ATCTCCATGAAGGCGCAATTCCACT
Phenotype
MIM: 605193 MIM: 615406
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIRAS3 PubMed Links

Gene Details

Gene
DIRAS3
Gene Name
DIRAS family GTPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004675.3 1820 Silent Mutation ATC,ATT I180I NP_004666.1
XM_017002773.1 1820 Silent Mutation ATC,ATT I180I XP_016858262.1
Gene
GNG12-AS1
Gene Name
GNG12 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details