Product Details

SNP ID
rs141181288
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:9038122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAGGTAGCTGGCCCCAGGACAG[A/C]GGCGTCTCCAGGACTCTTGGGCATG
Phenotype
MIM: 138230
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC2A5 PubMed Links

Gene Details

Gene
SLC2A5
Gene Name
solute carrier family 2 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135585.1 Intron NP_001129057.1
NM_003039.2 Intron NP_003030.1
XM_005263491.3 Intron XP_005263548.1
XM_017002133.1 Intron XP_016857622.1
XM_017002134.1 Intron XP_016857623.1
XM_017002135.1 Intron XP_016857624.1
XM_017002136.1 Intron XP_016857625.1
XM_017002137.1 Intron XP_016857626.1
XM_017002138.1 Intron XP_016857627.1
XM_017002139.1 Intron XP_016857628.1
XM_017002140.1 Intron XP_016857629.1
XM_017002141.1 Intron XP_016857630.1
XM_017002142.1 Intron XP_016857631.1

View Full Product Details