Product Details
- SNP ID
-
rs17040913
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:15256320 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGTCTCCTATAGAAATACAAGG[A/G]TTATCAAAAGTGAGTATTGACAATT
- Phenotype
-
MIM: 606400
MIM: 605612
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CAPN7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116552208] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAPN7
- Gene Name
- calpain 7
There are no transcripts associated with this gene.
- Gene
- SH3BP5
- Gene Name
- SH3 domain binding protein 5
- Gene
- SH3BP5-AS1
- Gene Name
- SH3BP5 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details