Product Details

SNP ID
rs189709984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37488032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCCTGTCTGTTTTAAGGGTAG[C/G]AGGAGCCATCTAGAGAGGTTTACTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POLR1E PubMed Links

Gene Details

Gene
POLR1E
Gene Name
RNA polymerase I subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282766.1 Intron NP_001269695.1
NM_022490.2 Intron NP_071935.1
XM_005251547.2 Intron XP_005251604.1

View Full Product Details