Product Details
- SNP ID
-
rs76572894
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:44622601 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGCAGCAAAGACAAAGAAATGAT[G/T]GCCTGTGCCAGTAGAAAAGAGATTA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
YIPF7
PubMed Links
Gene Details
- Gene
- YIPF7
- Gene Name
- Yip1 domain family member 7
View Full Product Details