Product Details

SNP ID
rs35311172
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:206932490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGTGCCGGGCTCTGGCCACCCC[C/G]ACAGCCACGGCTCCCACTGCCAGCA
Phenotype
MIM: 173880
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PIGR PubMed Links

Gene Details

Gene
PIGR
Gene Name
polymeric immunoglobulin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002644.3 2158 Silent Mutation GTC,GTG V658V NP_002635.2
XM_011509629.1 2158 Silent Mutation GTC,GTG V658V XP_011507931.1

View Full Product Details