Product Details

SNP ID
rs150918346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46468546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGTTCTTTCAACTCCTCAGTAT[C/G]GGTGGTCCCCCAGATCAAACCTTTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PNMAL1 PubMed Links

Gene Details

Gene
PNMAL1
Gene Name
paraneoplastic Ma antigen family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103149.1 1726 Missense Mutation CCA,CGA P377R NP_001096619.1
NM_018215.3 1726 UTR 3 NP_060685.2
XM_011527067.2 1726 Missense Mutation CCA,CGA P469R XP_011525369.1

View Full Product Details