Product Details
- SNP ID
-
rs4863506
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:139719550 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGGTATCTGCTGCTGTGCTGGGG[A/G]CTGGCTGAACGCTGGCATGCCTGGG
- Phenotype
-
MIM: 608991
MIM: 601733
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MAML3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61747885] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAML3
- Gene Name
- mastermind like transcriptional coactivator 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018717.4 |
3927 |
Missense Mutation |
CCC,TCC |
P1059S |
NP_061187.2 |
- Gene
- MGST2
- Gene Name
- microsomal glutathione S-transferase 2
View Full Product Details