Product Details

SNP ID
rs34059787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52126021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAATAGATGGCAACCTCCCTATC[C/T]GCCCGCAGGTCATAGAGGCGACACT
Phenotype
MIM: 604447
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNB5 PubMed Links

Gene Details

Gene
GNB5
Gene Name
G protein subunit beta 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006578.3 2519 Silent Mutation GCA,GCG A270A NP_006569.1
NM_016194.3 2519 Silent Mutation GCA,GCG A312A NP_057278.2
XM_011521162.2 2519 Silent Mutation GCA,GCG A270A XP_011519464.1
XM_011521163.2 2519 Silent Mutation GCA,GCG A218A XP_011519465.1
XM_017021867.1 2519 Silent Mutation GCA,GCG A129A XP_016877356.1

View Full Product Details