Product Details

SNP ID
rs60959366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:44624624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCAGTGAAAAGAACATGGCGCA[A/G]CCAGACAGGATGACCATGGGGAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YIPF7 PubMed Links

Gene Details

Gene
YIPF7
Gene Name
Yip1 domain family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182592.2 6015 Silent Mutation GGC,GGT G219G NP_872398.2
XM_011513679.2 6015 Silent Mutation GGC,GGT G257G XP_011511981.1

View Full Product Details