Product Details

SNP ID
rs3795966
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:201380521 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATCCTTCATGTGCTAACTTGTATAA[G/A]AGGTCAGTTAACTGCTGAACCTCAG
Phenotype
MIM: 607334
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
TRAK2 PubMed Links

Gene Details

Gene
TRAK2
Gene Name
trafficking kinesin protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015049.2 3117 UTR 3 NP_055864.2
XM_011511690.1 3117 UTR 3 XP_011509992.1
XM_017004772.1 3117 UTR 3 XP_016860261.1

View Full Product Details