Product Details

SNP ID
rs34599999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144432378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTAAGTGCAGGAGGGTGCCGGC[A/G]GGCAGGCTCTGCAGGGTCCTGGCCA
Phenotype
MIM: 604546 MIM: 611952
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6893 PubMed Links

Gene Details

Gene
MIR6893
Gene Name
microRNA 6893
There are no transcripts associated with this gene.

Gene
TONSL
Gene Name
tonsoku-like, DNA repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013432.4 3657 Silent Mutation CCC,CCT P1214P NP_038460.4
XM_011517048.2 3657 Silent Mutation CCC,CCT P890P XP_011515350.1
XM_011517049.2 3657 Silent Mutation CCC,CCT P878P XP_011515351.1
XM_011517050.2 3657 Intron XP_011515352.1
Gene
TONSL-AS1
Gene Name
TONSL antisense RNA 1
There are no transcripts associated with this gene.

Gene
VPS28
Gene Name
VPS28, ESCRT-I subunit
There are no transcripts associated with this gene.

View Full Product Details