Product Details

SNP ID
rs4758387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6107922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACCTCTTAGCTCTTGGTGCCAA[C/T]CTCCTCATCATAATCACCATTCAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR56B4 PubMed Links

Gene Details

Gene
OR56B4
Gene Name
olfactory receptor family 56 subfamily B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005181.2 239 Silent Mutation AAC,AAT N48N NP_001005181.1

View Full Product Details